Learn More
CRISPR-Cas9 Antibody, R&D Systems™
GREENER_CHOICE
Code produit 18762687
Change view
Click to view available options
Quantité:
100 μg
25 μg
Conditionnement:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18762687 25 μg 25µg
18734337 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18762687 Fournisseur R&D Systems Code fournisseur MAB10252SP
Mouse Monoclonal Antibody
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Multi-Species samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY
Spécification
Antigène CRISPR-Cas9
Applications Western Blot
Classification Monoclonal
Clone 1013816
Conjugué Unconjugated
Dilution Western Blot 2 μg/mL, Simple Western 20 μg/mL
Formule Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Numéro d’ordre du gène Q99ZW2
Alias de gène Cas9, CRISPR, CRISPR/Cas9, CRISPR-associated endonuclease Cas9/Csn1, CRISPR-associated protein 9 nuclease, CRISPR-Cas9/Csn1, csn1, SPy_1046, SPy1046, SpyCas9
Espèces hôtes Mouse
Immunogène E. coli-derived Cas9, Asp2-Asp1368, Accession # Q99ZW2
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 25 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 901176
Spécificité du test Detects Cas9 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Espèces cibles Multi-species
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Forme Purified
Isotype IgG2b