Learn More
Novus Biologicals™ CYB5R3 Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
0,5 mg
100 μg
Conditionnement:
0.50mg
100µg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 27-301 of Human CYB5R3 The Recombinant Human CYB5R3 Protein is derived from E. coli. The Recombinant Human CYB5R3 Protein has been validated for the following applications: SDS-Page.
Spécification
Spécification
| Concentration | 1 mg/mL |
| À utiliser avec (application) | SDS-PAGE |
| Formule | Liquid. In 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1 mM DTT, 0.1 M NaCl. |
| Poids moléculaire | 34 kDa |
| Méthode de purification | SDS-PAGE |
| Quantité | 0,5 mg |
| Source | Human |
| Conditions de stockage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| État réglementaire | RUO |
| Nom usuel | CYB5R3 |
| Afficher plus |