Learn More
ACROBiosystems Cynomolgus NKp46 / NCR1 / CD335 Protein, His Tag (MALS verified)
Marque: ACROBiosystems NC1-C52H4-1mg
Informations supplémentaires : Poids : 0.00010kg
Description
Natural cytotoxicity triggering receptor 1 (NCR1) is also known as Natural killer cell p46-related protein (NK-p46), Lymphocyte antigen 94 homolog (LY94), CD antigen CD335, which belongs to the natural cytotoxicity receptor (NCR) family. NCR1 contains two Ig-like (immunoglobulin-like) domains. NCR1 interacts with CD247 and FCER1G. NCR1 / CD335 may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.6 kDa. The protein migrates as 34-40 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001271509.1 | |
1 mg | |
Pro 22 - Arg 257 | |
NCR1,LY94,CD335,NK-p46,hNKp46 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
28.6 kDa | |
Cynomolgus NKp46, His Tag (NC1-C52H4) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Accession # Q95JB9-1). | |
-20°C | |
NKp46 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers & CD Antigens |