Learn More
ACROBiosystems Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, His Tag
Marque: ACROBiosystems BCA-C52H7-100UG
Informations supplémentaires : Poids : 0.00001kg
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.9 kDa. The protein migrates as 14-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
XP_005591343.1 | |
100 μg | |
Met 1 - Ala 53 | |
TNFRSF17,CD269,BCM,BCMA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
7.9 kDa | |
Cynomolgus / Rhesus macaque BCMA, His Tag (BCA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus. | |
-20°C | |
BCMA | |
Cynomolgus Monkey, Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |