missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus / Rhesus macaque CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
423.38€ - 3108.82€
Spécification
Numéro d’adhésion | XP_005569963.1 |
---|---|
Poids moléculaire | 46.2 kDa |
Source | Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-C5259) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Arg 191 (Accession # XP_005569963.1). In the region Ala 20 - Arg 191, the AA sequence of Cynomolgus and Rhesus macaque CD27 are homologus. |
Immunogène | Ala 20 - Arg 191 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876247
|
ACROBiosystems
CD7-C5259-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16417319
|
ACROBiosystems
CD7-C5259-1MG |
1 mg |
3108.82€
1mg |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CD Antigen CD27 is also known as Tumor necrosis factor receptor superfamily member 7 (TNFRSF7), which belongs to TNF-receptor superfamily. CD27 / TNFRSF7 is found in most T-lymphocytes. CD27 / TNFRSF7 is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. This receptor transduces signals that lead to the activation of NF-κB and MAPK8/JNK. CD27 has been shown to interact with SIVA1, TRAF2 and TRAF3.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.2 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
XP_005569963.1 | |
Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-C5259) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Arg 191 (Accession # XP_005569963.1). In the region Ala 20 - Arg 191, the AA sequence of Cynomolgus and Rhesus macaque CD27 are homologus. | |
-20°C | |
CD27 | |
Cynomolgus Monkey, Rhesus Macaque | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
46.2 kDa | |
Ala 20 - Arg 191 | |
CD27,TNFRSF7,S152,T14,Tp55 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |