missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP2F1 Polyclonal Antibody

Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Rhesus Monkey (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CYP2F1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to dehydrogenate 3-methylindole, an endogenous toxin derived from the fermentation of tryptophan, as well as xenobiotic substrates such as naphthalene and ethoxycoumarin.
Spécification
Spécification
| Antigène | CYP2F1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.69 mg/mL |
| Conjugué | Unconjugated |
| Formule | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Expression | CYP2F1 |
| Numéro d’ordre du gène | P24903 |
| Alias de gène | C2F1; CYP2F; Cyp2f1; Cyp2f2; Cyp2f4; CYP4502F4; CYPIIF1; CYPIIF2; Cytochrome P450 2F1; cytochrome P450 2F2; cytochrome P450 family 2 subfamily F member 1; cytochrome P450, family 2, subfamily F, polypeptide 1; cytochrome P450, family 2, subfamily f, polypeptide 2; cytochrome P450, family 2, subfamily f, polypeptide 4; cytochrome P450, subfamily 2F, polypeptide 1; cytochrome P450, subfamily IIF, polypeptide 1; cytochrome P450-NAH-2; flavoprotein-linked monooxygenase; microsomal monooxygenase; naphthalene dehydrogenase; Naphthalene hydroxylase; P450-NAH-2; xenobiotic monooxygenase |
| Symboles de gène(s) | CYP2F1 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?