missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCTN6 Polyclonal Antibody

Description
The antibody detects endogenous levels of total DCTN6 protein.
The protein encoded by this gene contains an RGD (Arg-Gly-Asp) motif in the N-terminal region, which confers adhesive properties to macromolecular proteins like fibronectin. It shares a high degree of sequence similarity with the mouse homolog, which has been suggested to play a role in mitochondrial biogenesis. The exact biological function of this gene is not known. [provided by RefSeq, Jul 2008].
Specifications
Specifications
| Antigène | DCTN6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.6 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Expression | DCTN6 |
| Numéro d’ordre du gène | O00399 |
| Alias de gène | AU044699; Dctn6; dynactin 6; dynactin subunit 6; Dynactin subunit p27; novel RGD-containing protein; p27; p27 dynactin subunit; protein WS-3; RGD containing protein; WS3; WS-3 |
| Symboles de gène(s) | DCTN6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?