Learn More
DCTN6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Code produit 16816744 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16816744 20 μL 20µL
16806744 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16816744 Fournisseur Proteintech Code fournisseur 169471AP20UL
Rabbit Polyclonal Antibody
The protein encoded by this gene contains an RGD (Arg-Gly-Asp) motif in the N-terminal region, which confers adhesive properties to macromolecular proteins like fibronectin. It shares a high degree of sequence similarity with the mouse homolog, which has been suggested to play a role in mitochondrial biogenesis. The exact biological function of this gene is not known.
Spécification
Antigène DCTN6
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Expression DCTN6
Numéro d’ordre du gène O00399, Q9WUB4
Alias de gène DCTN6, dynactin 6, Dynactin subunit 6, Dynactin subunit p27, Protein WS 3, WS 3, WS3
Symboles de gène(s) Dctn6
Espèces hôtes Rabbit
Immunogène DCTN6 Fusion Protein Ag10561
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 10671, 22428, 290798
Espèces cibles Rat, Mouse, Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG