missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an ATPase activated by a variety of RNA species, but not by dsDNA. This protein, and that encoded by DDX5 gene, are more closely related to each other than to any other member of the DEAD box family. This gene can encode multiple isoforms due to both alternative splicing and the use of alternative translation initiation codons, including a non-AUG (CUG) start codon.
Spécification
Spécification
| Antigène | DDX17 |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.32 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Expression | DDX17 |
| Numéro d’ordre du gène | Q501J6, Q92841 |
| Alias de gène | DDX17, DEAD box protein 17, DEAD box protein p72, P72, RH70, RNA dependent helicase p72 |
| Symboles de gène(s) | Ddx17 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?