Learn More
Dermatopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Code produit 16836983 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16836983 20 μL 20µL
16826983 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16836983 Fournisseur Proteintech Code fournisseur 105371AP20UL
Rabbit Polyclonal Antibody
Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.
Spécification
Antigène Dermatopontin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Expression DPT
Numéro d’ordre du gène Q07507, Q9QZZ6
Alias de gène Dermatopontin, DPT, TRAMP
Symboles de gène(s) Dpt
Espèces hôtes Rabbit
Immunogène Dermatopontin Fusion Protein Ag0801
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 1805, 289178, 56429
Espèces cibles Rat, Human, Mouse
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG