Learn More
Invitrogen™ Digoxin Monoclonal Antibody (DG D8-9), Invitrogen™
Code produit 17820476
Modifier l'affichage
Click to view available options
Quantité:
1 mg
100 μg
500 μg
Conditionnement:
100µg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17820476 1 mg 1mg
17880466 100 μg 100µg
11513423 500 μg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 17820476 Fournisseur Invitrogen™ Code fournisseur MA546171
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
MA1-10809 detects Digoxin. MA1-10809 has been successfully used in ELISA applications. The MA1-10809 immunogen is Digoxin, conjugated to KLH.
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Digoxin
Applications ELISA
Classification Monoclonal
Clone DG D8-9
Concentration 0.80 mg/mL
Conjugué Unconjugated
Formule PBS with no preservative; pH 7.4
Alias de gène C41H64O14; DGX; Lanoxin
Espèces hôtes Mouse
Immunogène Digoxin, conjugated to KLH.
Méthode de purification Protein G
Quantité 1 mg
État réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Chemical
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG1
Molécule cible Chemical