Learn More
Thermo Scientific™ Cartes IR jetables, 5 x 10 cm, fenêtre Kcl, 9,5 mm d’ouverture
Fournissent un substrat non absorbant monté de façon pratique destiné à l’analyse qualitative par infrarouge.
Marca: Thermo Scientific™ 043934.KJ
Code nomenclature Nacres: SB.22
Dettagli aggiuntivi : Poids : 0.01000kg
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Taille : 5 x 10 cm
- Fenêtre KCl
- Ouverture de 9,5 mm
Specifica
Disposable IR Cards | |
5 Pk. |