Learn More
DLL3 Rabbit, anti-Human, CoraLite Plus 647, Polyclonal, Proteintech
Code produit 18064995 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
100 μL
20 μL
Conditionnement:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18064995 20 μL 20µL
18079415 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18064995 Fournisseur Proteintech Code fournisseur CL6472553520UL
Rabbit Polyclonal Antibody
The DLL3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DLL3. This antibody recognizes human, mouse antigen. The DLL3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 647.
Spécification
Antigène DLL3
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugué CoraLite Plus 647
Symboles de gène(s) delta like 3 (Drosophila), Delta like protein 3, Delta3, DLL3, Drosophila Delta homolog 3, SCDO1
Espèces hôtes Rabbit
Quantité 20 μL
Primaire ou secondaire Primary
Identification génétique (Entrez) 10683
Espèces cibles Human, Mouse
Contenu et stockage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Poids moléculaire des anticorps 150 kDa
Forme Liquid
Isotype IgG