Learn More
Juchheim Laborgeraete™ Double Bosshead
High quality double socket made of malleable cast iron.
Marque:  Juchheim Laborgeraete™ 04810 S
Informations supplémentaires : Poids : 0.10000kg
Code produit. 10179110
  • 24.10€ / 1 pièce
Expédition estimée: 17-06-2024
Description
  • Durable due to robust malleable cast iron
  • Option of safety screws for fixing by hand or lobe screws for mounting by hand
  • Made in Germany
Spécification
Spécification
Tempered Cast Iron
30 mm
Safety screws for fixing by hand