Learn More
Juchheim Laborgeraete™ Double Bosshead
High quality double socket made of malleable cast iron.
Marque: Juchheim Laborgeraete™ 04820 S
Informations supplémentaires : Poids : 0.10000kg
Description
- Durable due to robust malleable cast iron
- Option of safety screws for fixing by hand or lobe screws for mounting by hand
- Made in Germany
Spécification
Tempered Cast Iron | |
20 mm | |
Safety screws for fixing by hand | |
120 g |