missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPF1 Polyclonal Antibody

Description
Additional Information: This antibody is expected to recognize all three reported isoforms (NP_001128627.1; NP_004638.2; NP_001128628.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
May have an important role in developing neurons by participating in regulation of cell survival, possibly as a neurospecific transcription factor. Belongs to the neuron-specific chromatin remodeling complex (nBAF complex). During neural development a switch from a stem/progenitor to a post-mitotic chromatin remodeling mechanism occurs as neurons exit the cell cycle and become committed to their adult state. The transition from proliferating neural stem/progenitor cells to post-mitotic neurons requires a switch in subunit composition of the npBAF and nBAF complexes. As neural progenitors exit mitosis and differentiate into neurons, npBAF complexes which contain ACTL6A/BAF53A and PHF10/BAF45A, are exchanged for homologous alternative ACTL6B/BAF53B and DPF1/BAF45B or DPF3/BAF45C subunits in neuron-specific complexes (nBAF). The npBAF complex is essential for the self-renewal/proliferative capacity of the multipotent neural stem cells. The nBAF complex along with CREST plays a role regulating the activity of genes essential for dendrite growth.
Spécification
Spécification
| Antigène | DPF1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Expression | DPF1 |
| Numéro d’ordre du gène | Q92782 |
| Alias de gène | Baf45b; BRG1-associated factor 45B; D4, zinc and double PHD fingers family 1; double PHD fingers 1; DPF1; NEU BAF45b; NEUD4; neuro-d4; neuro-d4 homolog; neuronal d4 domain family member; zinc finger protein neuro-d4 |
| Symboles de gène(s) | DPF1 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?