missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF3l Polyclonal Antibody
GREENER_CHOICE

Code produit 13406177
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
13406177 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 13406177 Fournisseur Invitrogen™ Code fournisseur PA531647

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brown, white adipose. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (94%), Dog (100%), Pig (100%), Chicken (98%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis (PubMed:17581632, PubMed:25849773, PubMed:27462815). The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation (PubMed:17581632). The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression (PubMed:25849773). [UniProt].
TRUSTED_SUSTAINABILITY

Spécification

Antigène eIF3l
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 20% glycerol and 0.01% thimerosal; pH 7
Expression EIF3L
Numéro d’ordre du gène Q8QZY1, Q9Y262
Alias de gène 0610011H21Rik; 66 kDa tyrosine-rich heat shock protein; 67 kDa polymerase-associated factor; D15N1e; eIEF associated protein HSPC021; EIF3EIP; Eif3ip; EIF3L; EIF3S11; Eif3s6ip; Eukaryotic translation initiation factor 3 subunit 6-interacting protein; eukaryotic translation initiation factor 3 subunit E-interacting protein; Eukaryotic translation initiation factor 3 subunit L; eukaryotic translation initiation factor 3, subunit 6 interacting protein; eukaryotic translation initiation factor 3, subunit L; HSP-66Y; HSPC021; HSPC025; MSTP005; Paf67
Symboles de gène(s) EIF3L
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the C-terminus region of human EIF3L. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 223691, 51386
Espèces cibles Human, Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.