Learn More
Veolia Water Systems™ Cartouche Elgas™
Deionization cartridge compatible with systems that remove ionic contaminants to achieve specified water quality.
Marque: Veolia Water Systems™ LC141
Code nomenclature Nacres: JA.13
Informations supplémentaires : Poids : 2.16000kg
Description
- Removes ionic contaminants to achieve specified water quality
- Compatible with MEDICA-D 7/15, MEDICA-R 7/15, MEDICA-S 7/15,
- PURELAB Option-R 7/15, PURELAB Option-S 7/15
- Maximum operating temperature 95 degrees F