Learn More
3M™ Enrichment Pouch
A heavyweight, puncture-resistant pouch, ideal for many sample preparation and enrichment procedures.
176.00€
Code produit | Marque | Comprend | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Comprend | Prix | Quantité et disponibilité | |||||
17268520
|
3M™
7000030308 |
Bouillon d’eau peptoné tamponné de 225 ml |
176.00€
32 pièces |
Expédition estimée: 26-06-2024
Connectez-vous pour voir le stock disponible |
|||||
Description
- Gamma irradiated, ready-to-use pouch with 225 mL Buffered Peptone Water Broth requires no mixing or sterilizing.
- Flat bottom, stand-up pouch ensures stability when adding samples.
- Wire tie holds top open for easy sample addition and closes pouch for transportation.
- Multi-laminate polyester pouch is puncture resistant.
- 2 year shelf life from date of manufacture provides long-term readiness.
Spécification
32/Cs. |