missing translation for 'onlineSavingsMsg'
Learn More

PeproTech Eotaxin Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Code produit 17835063
Change view
Click to view available options
Quantité:
1 mg
100 μg
50 μg
Conditionnement:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17835063 1 mg 1mg
17805053 50 μg 50µg
17865803 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17835063 Fournisseur PeproTech Code fournisseur 500P411MG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Eotaxin (CCL11). Anti-Human Eotaxin (CCL11)-specific antibody was purified by affinity chromatography employing an immobilized Human Eotaxin (CCL11) matrix. Sandwich ELISA: To detect Human Eotaxin (CCL11) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Eotaxin (CCL11) (500-P41Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Eotaxin (CCL11). Western Blot: To detect hEotaxin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hEotaxin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
TRUSTED_SUSTAINABILITY

Spécification

Antigène Eotaxin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugué Unconjugated
Formule PBS with no preservative
Expression CCL11
Numéro d’ordre du gène P51671
Alias de gène C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11
Symboles de gène(s) CCL11
Espèces hôtes Rabbit
Immunogène E.coli-derived Recombinant Human Eotaxin (CCL11).
Méthode de purification Antigen affinity chromatography
Quantité 1 mg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 6356
Espèces cibles Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Lyophilized
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.