missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ERF Polyclonal Antibody
GREENER_CHOICE

Code produit 16331725
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16331725 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16331725 Fournisseur Invitrogen™ Code fournisseur PA584773

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Immunogen sequence: SSSSLFSAVV ARRLGRGSVS DCSDGTSELE EPLGEDPRAR PPGPPDLGAF RGPPLARLPH DPGVF Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).
TRUSTED_SUSTAINABILITY

Specifications

Antigène ERF
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugué Unconjugated
Formule PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Expression ERF
Numéro d’ordre du gène P50548
Alias de gène CRS4; ERF; Est2 repressor factor; ETS domain-containing transcription factor ERF; ETS domain-containing transcription factor ERF; ets2 repressor factor; Ets2 repressor factor; fc22h01; LOW QUALITY PROTEIN: ETS domain-containing transcription factor ERF; PE 2; PE2; PE-2; wu:fc22h01
Symboles de gène(s) ERF
Espèces hôtes Rabbit
Immunogène Recombinant protein corresponding to Human ERF. Recombinant protein control fragment (Product #RP-103876).
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 2077
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.