missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FHL1 Recombinant Rabbit Monoclonal Antibody (K01_2M41)
GREENER_CHOICE

Code produit 30829369
Change view
Click to view available options
Quantité:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30829369 100 μL
30826560 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30829369 Fournisseur Invitrogen™ Code fournisseur MA564064

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: AEKFDCHYCR DPLQGKKYVQ KDGHHCCLKC FDKFCANTCV ECRKPIGADS KEVHYKNRFW HDTCFRCAKC LHPLANETFV AKDNKILCNK CTTREDSPKC KGCFKAIVAG DQNVEYKGTV WHKDCFTCSN CKQVIGTGSF FPKGEDFYCV TCHETKFAKH CVKCNKAITS GGITYQDQPW HADCFVCVTC SKKLAGQRFT AVEDQYYCVD CYKNFVAKKC AGCKNPITGK RTVSRVSHPV SKARKPPVCH GKRLPLTLFP SANLRGRHPG GERTCPSWVV VLYRKNRSLA APRGPGLVKA PVWWPMKDNP GTTTASTAKN AP Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%, Pig - 98%, Cynomolgus monkey - 100%.

This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. Expression of these family members occurs in a cell- and tissue-specific mode and these proteins are involved in many cellular processes. Mutations in this gene have been found in patients with Emery-Dreifuss muscular dystrophy. Multiple alternately spliced transcript variants which encode different protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Spécification

Antigène FHL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2M41
Concentration 1 mg/mL
Conjugué Unconjugated
Formule 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Expression FHL1
Numéro d’ordre du gène Q13642
Alias de gène bA535K18.1; cb1039; fhl; Fhl1; FHL-1; FHL1A; fhl1b; fhla; FLH1A; four and a half LIM domains; four and a half LIM domains 1; four and a half LIM domains 1a; four and a half LIM domains 1b; four and a half LIM domains 1b; four and a half LIM domains protein 1b; four and a half LIM domains a; four and a half LIM domains protein 1; four and a half LIM domains protein 1; four and a half LIM domains protein 1a; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; KYOT; KYO-T; LIM protein; LIM protein SLIMMER; MGC111107; RAM14-1; RBMX1A; RBMX1B; RBP-associated molecule 14-1; RP11-535K18.1; skeletal muscle LIM-protein 1; SLIM; SLIM1; SLIM-1; SLIMMER; XMPMA; zgc:92025
Symboles de gène(s) FHL1
Espèces hôtes Rabbit
Immunogène Recombinant protein of human FHL1 (aa 2-323).
Méthode de purification Protein A/G
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 2273
Espèces cibles Human
Contenu et stockage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.