Learn More
Fisherbrand™ 35mm Silicone O-Ring
Ideal for high temperature applications in static conditions
Marque: Fisherbrand™
Informations supplémentaires : Poids : 0.05000kg
Spécification
O-Ring | |
Silicone | |
3.5 cm | |
1.3 in. |