missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FL - SRC 3-2 Peptide, 100μM
Fl - SRC 3-2 peptide, 100μM
Marque: Invitrogen PV4592
Code nomenclature Nacres: NA.26
Informations supplémentaires : Poids : 0.50000kg
Description
LanthaScreen™ fluorescent nuclear receptor coregulator peptides contain known interaction motifs and are labeled with fluorescein. These peptides are matched and validated to complement the LanthaScreen™ TR-FRET Nuclear Receptor Coregulator Assays. Assays developed using these reagents enable primary or secondary screening of nuclear receptor agonists and/or antagonists.How they work
Ligand binding to nuclear receptors causes conformational changes in the receptor, resulting in a cascade of events, including dissociation of repressor proteins, association of coactivator proteins, and assembly of pol II and other transcriptional factors for activation of target genes. TR-FRET based assays can be developed using the LanthaScreen™ panel of fluorescein-labeled coregulator peptides to investigate conformational changes of nuclear receptors upon ligand binding, either by determining the affinity of ligand-bound receptor for different coregulator peptides, or by identifying additional agonists or antagonists via displacement or recruitment of a specific coregulator peptide. In the LanthaScreen™ TR-FRET RXR beta Coactivator Assay, a terbium (Tb)-labeled anti-GST antibody is used to indirectly label GSTtagged RXR beta protein. An agonist (9-cis Retinoic Acid) added to the receptor which, upon ligand binding, causes a conformational change resulting in recruitment of the Fl-D22 coactivator peptide (Figure 1). The binding of the fluorescent coregulator peptide to RXR beta causes an increase in the TR-FRET emission ratio (Figure 2).
Contents and Storage:
LanthaScreen™ fluorescent nuclear receptor coregulator peptides are supplied as 100μM solutions in peptide-appropriate buffers. Store peptides at -20°C.
Spécification
≥96% by HPLC | |
100 μm | |
Fluorescein-SRC 3-2 Peptide, 100μL, 100μM Concentration, 2556Da Molecular Weight, –5 to –30°C Storage Condition, Used for Pharma and Biopharma, Target and Lead Identification and Validation | |
2556Da | |
100 μL | |
Fluorescein-LQEKHRILHKLLQNGNSPA |
Fluorescein - SRC 3-2 Peptide | |
-5°C to -30°C | |
Pharma and biopharma, target and lead identification and validation, drug discovery and development, nuclear receptor biology, nuclear receptor proteins, biochemical nuclear receptor assays, nuclear receptor coregulator interaction | |
Fluorescein - SRC 3-2 Peptide | |
For research use only. Not for use in diagnostic procedures | |
50mM HEPES (pH 7.5) |
For Research Use Only. Not for use in diagnostic procedures.