Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Wide Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marque:  Lenz Laborglasintrumente 07050028
Informations supplémentaires : Poids : 0.01000kg
Code produit. 11923831
  • 350.86€ / 10 pièces
Expédition estimée: 27-06-2024
Description
  • Acc. to DIN ISO 24450
  • With beaded rim
  • Material: DURAN™
Spécification
Spécification
50 mL
Duran Glass
90 mm