Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Wide Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marque: Lenz Laborglasintrumente 07050070
Informations supplémentaires : Poids : 0.01000kg
Description
- Conforme à la norme DIN ISO 24450
- Avec rebord perlé
- Matériau : DURAN™
Spécification
1 L | |
Verre Duran | |
190 mm |