Learn More
Invitrogen™ FLRT1 Monoclonal Antibody (357721)
Code produit 30535386
Change view
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30535386 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30535386 Fournisseur Invitrogen™ Code fournisseur MA561205
Mouse Monoclonal Antibody
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. The family members may function in cell adhesion and/or receptor signalling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix. The encoded protein shares sequence similarity with two other family members, FLRT2 and FLRT3. This gene is expressed in kidney and brain.
TRUSTED_SUSTAINABILITY
Spécification
Antigène FLRT1
Applications Western Blot
Classification Monoclonal
Clone 357721
Concentration 0.5 mg/mL
Conjugué Unconjugated
Formule PBS with trehalose and no preservative
Expression FLRT1
Numéro d’ordre du gène Q9NZU1
Alias de gène AW742165; D630040I23Rik; Fibronectin leucine rich transmembrane protein 1; fibronectin leucine rich transmembrane protein 3; fibronectin-like domain-containing leucine-rich transmembrane protein 1; Flrt1; Leucine-rich repeat transmembrane protein FLRT1; RGD1565152; SPG68; UNQ752/PRO1483
Symboles de gène(s) FLRT1
Espèces hôtes Mouse
Immunogène Mouse myeloma cell line NS0-derived recombinant human FLRT1, Ile21-Pro524.
Méthode de purification Protein A/G
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 23769
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Lyophilized
Isotype IgG2b