Description
May catalyze alpha-1,3 glycosidic linkages involved in the expression of sialyl Lewis X antigens.
Specifications
Specifications
| Antigène | FUT7 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.47 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Expression | FUT7 |
| Numéro d’ordre du gène | Q11130, Q11131 |
| Alias de gène | Alpha (1,3) fucosyltransferase, Fuc TVII, Fucosyltransferase 7, Fucosyltransferase VII, FucT VII, FUT7, Selectin ligand synthase |
| Symboles de gène(s) | Fut7 |
| Show More |