Learn More
Karl Hecht™ Petri Dish, Glass
Shallow, lidded glass dishes for culturing cells including bacteria. Karl Hecht™ Petri Dishes, Glass are available in a range of convenient sizes.
Marque:  Karl Hecht™ 41042042
Code nomenclature Nacres: NB.13
Informations supplémentaires : Poids : 0.50000kg
Code produit. 10257281
  • 21.15€ / 1 pièce
Expédition estimée: 18-07-2024
Description
  • Glass construction.
  • Lidded.
  • Available in a variety of sizes.
Spécification
Spécification
Glass
Round
5 cm
20 cm