missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLMN Polyclonal Antibody

Description
Recommended positive controls: Jurkat, Raji, NIH-3T3. Predicted reactivity: Mouse (85%), Rat (84%), Chimpanzee (99%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a phosphorylated protein that is a member of a Skp1-Cullin-F-box-like complex. The protein is essential for normal development of the vasculature and mutations in this gene have been associated with glomuvenous malformations, also called glomangiomas. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
Spécification
Spécification
| Antigène | GLMN |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Expression | GLMN |
| Numéro d’ordre du gène | Q8BZM1, Q92990 |
| Alias de gène | 9330160J16Rik; AW227515; FAP; Fap48; FAP68; FK506-binding protein-associated protein; FKBP associated protein; FKBPAP; FKBP-associated protein; GLML; Glmn; Glomulin; glomulin, FKBP associated protein; glomulin-like protein; GVM; LOW QUALITY PROTEIN: glomulin; VMGLOM |
| Symboles de gène(s) | GLMN |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?