missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLP2R Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset (100%) Gibbon (94%) Bovine, Dog, Elephant, Panda, Pig (89%) Bat, Rabbit (83%).
The GLP2 receptor (GLP2R) is a G protein-coupled receptor superfamily member closely related to the glucagon receptor ans GLP1 receptor. Glucagon-like peptide-2 (GLP2) is a 33-amino acid proglucagon-derived peptide produced by intestinal enteroendocrine cells. Like glucagon-like peptide-1 (GLP1) and glucagon itself, it is derived from the proglucagon peptide encoded by the GCG gene. GLP2 stimulates intestinal growth and upregulates villus height in the small intestine, concomitant with increased crypt cell proliferation and decreased enterocyte apoptosis. Moreover, GLP2 prevents intestinal hypoplasia resulting from total parenteral nutrition. GLP2R, a G protein-coupled receptor superfamily member is expressed in the gut and closely related to the glucagon receptor (GCGR) and the receptor for GLP1 (GLP1R).
Spécification
Spécification
| Antigène | GLP2R |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.1% sodium azide |
| Expression | GLP2R |
| Numéro d’ordre du gène | O95838 |
| Alias de gène | 9530092J08Rik; G protein-coupled receptor GLP2R; GLP-2; Glp2 receptor; GLP-2 receptor; GLP2R; GLP-2R; GLP-2-R; glucagon like peptide 2 receptor; glucagon-like peptide 2 receptor; glucagon-like peptide-2 receptor |
| Symboles de gène(s) | GLP2R |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?