missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GM-CSF Monoclonal Antibody (1089)

Code produit 11854281
Change view
Click to view available options
Quantité:
500 μg
Conditionnement:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11854281 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11854281 Fournisseur Invitrogen™ Code fournisseur M501

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

M501 targets Human GM-CSF in ELISA applications and shows reactivity with Human samples. The M501 immunogen is glycosylated, CHO-derived recombinant human GM-CSF. M501 detects Human GM-CSF which has a predicted molecular weight of approximately 14 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).

The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
TRUSTED_SUSTAINABILITY

Spécification

Antigène GM-CSF
Applications ELISA
Classification Monoclonal
Clone 1089
Concentration 1.0 mg/mL
Conjugué Unconjugated
Formule PBS with no preservative
Expression CSF2
Numéro d’ordre du gène P04141
Alias de gène colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Symboles de gène(s) CSF2
Espèces hôtes Mouse
Immunogène Glycosylated, CHO-derived recombinant human GM-CSF.
Méthode de purification Protein A
Quantité 500 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 1437
Espèces cibles Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.