Learn More
CCDC50, Goat, Polyclonal Antibody, Abnova™
Description
This gene encodes a soluble, cytoplasmic, tyrosine-phosphorylated protein with multiple ubiquitin-interacting domains. Mutations in this gene cause nonsyndromic, postlingual, progressive sensorineural DFNA44 hearing loss. In mouse, the protein is expressed in the inner ear during development and postnatal maturation and associates with microtubule-based structures. This protein may also function as a negative regulator of NFkb signaling and as an effector of epidermal growth factor (EGF)-mediated cell signaling. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Spécification
Spécification
| Antigène | CCDC50 |
| Applications | ELISA, Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Description | Goat polyclonal antibody raised against synthetic peptide of CCDC50. |
| Dilution | ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. |
| Formule | In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) |
| Expression | CCDC50 |
| Numéro d’ordre du gène | NP_848018.1;NP_777568.1 |
| Afficher plus |
For Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.