missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GP1BB Polyclonal Antibody

Description
Recommended positive controls: Raji. Predicted reactivity: Mouse (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD42a is a single-chain membrane glycoprotein that forms a noncovalent complex with CD42b. CD42b, also known as glycoprotein Ib a (GPIb a) is a membrane glycoprotein that is composed of a and b chains. The CD42b b chain is also designated CD42c, and is expressed on platelets and megakaryoctes. CD42a and CD42b are also present on platelets and mega-karyocytes, and the complex is a major component of the platelet surface. The complex acts as a receptor for von Willebrand's factor and as a von Willebrand's factor-dependent adhesion receptor.
Spécification
Spécification
| Antigène | GP1BB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.75 mg/mL |
| Conjugué | Unconjugated |
| Formule | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Expression | GP1BB |
| Numéro d’ordre du gène | P13224, P56400 |
| Alias de gène | Antigen CD42b-beta; BDPLT1; BS; CD42c; glycoprotein Ib (platelet), beta polypeptide; glycoprotein Ib platelet beta subunit; glycoprotein Ib, beta polypeptide; Gp1bb; GP-Ib beta; GPIbB; GPIbbeta; GPIb-beta; nuclear localization signal deleted in velocardiofacial syndrome; platelet glycoprotein Ib beta chain; platelet membrane glycoprotein Ib beta; truncated platelet membrane glycoprotein Ib beta |
| Symboles de gène(s) | GP1BB |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?