missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1 (Ly-6G/Ly-6C) Rat, anti-Mouse, PE, Clone: RB6-8C5, STEMCELL Technologies
Description
- The RB6-8C5 antibody reacts with the structurally related murine Ly-6G (21 - 25 kDa) and Ly-6C (14 - 16 kDa) GPI-anchored proteins, which together comprise the granulocyte receptor-1 antigen (Gr-1)
- Gr-1 is expressed on monocytes, neutrophils, subsets of macrophages, plasmacytoid dendritic cells, and T cells
- Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
- In the periphery Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
- Whereas the RB6-8C5 binds to both Ly-6G and Ly-6C, another commonly used anti-Gr-1 antibody, clone 1A8, binds specifically to Ly-6G
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications
Specifications
| Antigène | Gr-1 (Ly-6G/Ly-6C) |
| Applications | Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | RB6-8C5 |
| Conjugué | PE |
| Espèces hôtes | Rat |
| Quantité | 50 μg |
| Primaire ou secondaire | Primary |
| Espèces cibles | Mouse |
| Isotype | IgG2b κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?