missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GRP Protein

Code produit. 18588303 Tous les produits Bio Techne Produits
Click to view available options
Quantité:
0,5 mg
Conditionnement:
0.50mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 18588303

Marque: Novus Biologicals™ NBP1724450.5mg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-172 of Human GRP The Recombinant Human GRP Protein is derived from E. coli. The Recombinant Human GRP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

À utiliser avec (application) SDS-PAGE
Formule Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl.
Poids moléculaire 21.6kDa
Méthode de purification SDS-PAGE
Quantité 0,5 mg
Source Human
Conditions de stockage Store at -80°C. Avoid freeze-thaw cycles.
État réglementaire RUO
Nom usuel GRP
Conjugué Unconjugated
Pureté ou qualité >85%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.