Learn More
Guinea Pig anti-Rabbit IgG (H+L) Secondary Antibody, PE/Cy5.5 (Pre-adsorbed), Novus Biologicals™
Code produit 18545612 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
500 μg
Conditionnement:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18545612 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18545612 Fournisseur Novus Biologicals Code fournisseur NBP172763PECY55
Please to purchase this item. Need a web account? Register with us today!
Guinea Pig Polyclonal Antibody
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Spécification
Antigène IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugué PE-Cyanine5.5
Formule PBS
Espèces hôtes Guinea Pig
Immunogène Rabbit IgG whole molecule
Méthode de purification Multi-step
Quantité 500 μg
État réglementaire RUO
Disciplines de recherche Adaptive Immunity, Immunology
Primaire ou secondaire Secondary
Espèces cibles Rabbit
Contenu et stockage Store at 4°C in the dark. Do not freeze.
Isotype IgG