missing translation for 'onlineSavingsMsg'
Learn More

H2AFY2 Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene

Product Code. 17592413
Encompass_Preferred
Change view
Click to view available options
Quantité:
30 μL
100 μL
Unit Size:
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
17592413 30 μL 30µL
17572413 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17592413 Supplier Origene Technologies Supplier No. TA803386S

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

H2AFY2 Monoclonal antibody specifically detects H2AFY2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigène H2AFY2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C2
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Expression ZNF286A
Numéro d’ordre du gène Q9HBT8
Alias de gène Core histone macro-H2A.2; core histone macroH2A2.2; H2A histone family member Y2; H2A histone family, member Y2; H2AFY2; H2afy3; Histone macroH2A2; histone mH2A2; macroH2A2; mH2A2; RGD1561371
Espèces hôtes Mouse
Immunogène Human recombinant protein fragment corresponding to amino acids 1-252 of human ZNF286A produced in E.coli.
Méthode de purification Affinity chromatography
Quantité 30 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 57335
Espèces cibles Human
Contenu et stockage -20° C, Avoid Freeze/Thaw Cycles
Forme Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.