missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HABP2 Polyclonal Antibody

Description
Recommended positive controls: 4549. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HABP2 is an extracellularserine protease that binds hyaluronic acid and is involved in celladhesion. The encoded protein is synthesized as a single chain, butthen undergoes an autoproteolytic event to form the functionalheterodimer. Further autoproteolysis leads to smaller, inactivepeptides. This protease is known to cleave urinary plasminogenactivator, coagulation factor VII, and the alpha and beta chains offibrinogen, but not prothrombin, plasminogen, or the gamma chain offibrinogen. Two transcript variants encoding different isoformshave been found for this gene.
Spécification
Spécification
| Antigène | HABP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Expression | HABP2 |
| Numéro d’ordre du gène | Q14520 |
| Alias de gène | AI035669; Factor seven-activating protease; factor VII activating protein; factor VII-activating protease; FSAP; HABP; Habp2; Hepatocyte growth factor activator-like protein; HGFAL; hyaduronic acid-binding protein 2; hyaluronan binding protein 2; hyaluronan-binding protein 2; Hyaluronan-binding protein 2 27 kDa light chain; Hyaluronan-binding protein 2 27 kDa light chain alternate form; Hyaluronan-binding protein 2 50 kDa heavy chain; Hyaluronan-binding protein 2 50 kDa heavy chain alternate form; hyaluronic acid binding protein 2; NMTC5; PHBP; plasma hyalurona; plasma hyaluronan binding protein; plasma hyaluronan-binding protein |
| Symboles de gène(s) | HABP2 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?