missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ hnRNP A1 Protein

Product Code. 18442991 Shop All Bio Techne Products
Click to view available options
Quantité:
0,5 mg
100 μg
Unit Size:
0.50mg
100µg
This item is not returnable. View return policy

Product Code. 18442991

Brand: Novus Biologicals™ NBP1991060.5mg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-320 of Human hnRNP A1 The Recombinant Human hnRNP A1 Protein is derived from E. coli. The Recombinant Human hnRNP A1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
À utiliser avec (application) SDS-PAGE
Formule Liquid. 20 mM Tris-HCl buffer (pH 8.0) containing 40% glycerol, 0.15 M NaCl, 1 mM DTT
Poids moléculaire 36.6 kDa
Méthode de purification SDS-PAGE
Quantité 0,5 mg
Source Human
Conditions de stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
État réglementaire RUO
Nom usuel hnRNP A1
Conjugué Unconjugated
Pureté ou qualité >85%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.