Learn More
HO-1/HMOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Code produit 16867253 Tous les produits Proteintech Produits
Modifier l'affichage
Click to view available options
Quantité:
150 μL
20 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16867253 150 μL 150µL
16877253 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16867253 Fournisseur Proteintech Code fournisseur 107011AP150UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
Spécification
Antigène HO-1/HMOX1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Expression HMOX1
Numéro d’ordre du gène P06762, P09601, P14901
Alias de gène bK286B10, heme oxygenase (decycling) 1, Heme oxygenase 1, HMOX1, HO, HO 1, HO1, HSP32
Symboles de gène(s) HMOX1
Espèces hôtes Rabbit
Immunogène HO-1/HMOX1 Fusion Protein Ag1190
Méthode de purification Antigen Affinity Chromatography
Quantité 150 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 15368, 24451, 3162
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG