Learn More
Juchheim Laborgeraete™ Hook Bosshead
Hook socket for right-angled connection of tripod rods.
Marque:  Juchheim Laborgeraete™ 04860
Informations supplémentaires : Poids : 0.50000kg
Code produit. 10538841
  • 11.90€ / 1 pièce
Expédition estimée: 26-06-2024
Description
  • With prism to precise 90° arrangement of two tripod rods
  • High quality and durable design
  • Made in Germany
Spécification
Spécification
Steel
15 mm