missing translation for 'onlineSavingsMsg'
Learn More

HOXD10 Mouse anti-Human, Clone: OTI1D11, lyophilized, TrueMAB™

Code produit 17656301
Encompass_Preferred
Change view
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
17656301 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17656301 Fournisseur Origene Technologies Code fournisseur CF800777

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm′s tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.

Spécification

Antigène HOXD10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D11
Conjugué Unconjugated
Formule PBS with 8% trehalose and no preservative; pH 7.3
Expression HOXD10
Numéro d’ordre du gène P28358
Alias de gène HOX4, HOX4D, HOX4E, Hox-4.4
Symboles de gène(s) HOXD10
Espèces hôtes Mouse
Immunogène Human recombit protein fragment corresponding to amino acids 80-340 of human HOXD10 produced in E.coli.
Méthode de purification Affinity Chromatography
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 3236
Espèces cibles Human
Contenu et stockage -20° C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Lyophilized
Isotype IgG2b
Afficher plus Afficher moins
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.