Learn More
ACROBiosystems HRSV (A) Glycoprotein G Protein, His Tag
423.38€ - 2153.18€
Spécification
Poids moléculaire | 27.0 kDa |
---|---|
Source | HRSV (A) Glycoprotein G, His Tag (RSG-V5221) is expressed from human 293 cells (HEK293). It contains AA His 67 - Arg 297 (Accession # P20895). |
Immunogène | His 67 - Arg 297 |
Conditions de stockage | -20°C |
Alias de gène | Glycoprotein G,G,mG,Attachment glycoprotein G,Membrane-bound glycoprotein |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15877627
|
ACROBiosystems
RSG-V5221-50UG |
50 μg |
423.38€
50µg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867627
|
ACROBiosystems
RSG-V5221-1MG |
1 mg |
2153.18€
1mg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Human respiratory syncytial virus (HRSV) is the most common etiological agent of acute lower respiratory tract disease in infants and can cause repeated infections throughout life. Human respiratory syncytial virus A (strain Long) major surface glycoprotein G (RSV-G), a member of the pneumoviruses glycoprotein G family, is also known as attachment glycoprotein G and membrane-bound glycoprotein (mG), which contains a linear heparin binding domain essential for virus attachment to the host. Concretely speaking, RSV-G can attache the virion to the host cell membrane by interacting with heparan sulfate, initiating the infection. Furthermore, RSV-G can also interact with host CX3CR1, the receptor for the CX3C chemokine fractalkine, to modulate the immune response and facilitate infection. Unlike the other paramyxovirus attachment proteins, RSV-G lacks both neuraminidase and hemagglutinating activities.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.0 kDa. The protein migrates as 46-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
27.0 kDa | |
His 67 - Arg 297 | |
Glycoprotein G,G,mG,Attachment glycoprotein G,Membrane-bound glycoprotein | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
HRSV (A) Glycoprotein G, His Tag (RSG-V5221) is expressed from human 293 cells (HEK293). It contains AA His 67 - Arg 297 (Accession # P20895). | |
-20°C | |
Glycoprotein G / G | |
HRSV | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |