Learn More
HSP90 Mouse anti-Human, CoraLite 594, Clone: 3F11C1, Proteintech
Tous les produits Proteintech ProduitsDescription
The HSP90 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSP90. This antibody recognizes human, mouse, rat, pig antigen. The HSP90 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spécification
Spécification
| Antigène | HSP90 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 3F11C1 |
| Concentration | 2000 μg/mL |
| Conjugué | CoraLite 594 |
| Espèces hôtes | Mouse |
| Quantité | 100 μL |
| Primaire ou secondaire | Primary |
| Identification génétique (Entrez) | 3320 |
| Afficher plus |