missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPBAP1 Polyclonal Antibody

Description
Peptide sequence: SDGKDFVDKD GEHFGKLHCA KRQQIMSNSE NAIEEQIASN TTTTPQTFIS Sequence homology: Human: 100%.
HSPBAP1 (HSPB (heat shock 27kDa) associated protein 1), also known as PASS1 (protein associated with small stress protein 1), is a 488 amino acid cytoplasmic protein that contains one JMJC (Jumonji C) domain and shares 80% identity with its rat homolog, PASS1. Widely expressed with highest expression in ovary, thymus and pancreas, HSPBAP1 is thought to play a role in mediating cellular stress responses within the cell. Due to the presence of a JMJC domain, HSPBAP1 may be involved in chromatin remodeling events. Defects or translocations in the gene encoding HSPBAP1 are associated with renal cell carcinoma 1 (RCC1), suggesting a possible role for HSPBAP1 in carcinogenesis. Three isoforms of HSPBAP1 exist due to alternative splicing events.
Spécification
Spécification
| Antigène | HSPBAP1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 2% sucrose and 0.09% sodium azide |
| Expression | HSPBAP1 |
| Numéro d’ordre du gène | Q96EW2 |
| Alias de gène | 27 kDa heat shock protein-associated protein 1; 3830421G21Rik; FLJ22623; FLJ39386; Heat shock protein; HSP; HSP27; HSP28; HSPB (heat shock 27kDa) associated protein 1; Hspb associated protein 1; HSPB1 associated protein 1; HSPB1-associated protein 1; HSPBAP1; PASS1; protein associated with small stress protein 1; protein associating with small stress protein PASS1 |
| Symboles de gène(s) | HSPBAP1 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?