Learn More
Human Bcl-w Antibody, R&D Systems™
Code produit 18139440
Modifier l'affichage
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18139440 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18139440 Fournisseur R&D Systems Code fournisseur MAB824
 Article restreint
Expédition estimée: 25-11-2026
Mouse Monoclonal Antibody
BCL-W/BCL2L2 Monoclonal specifically detects BCL-W/BCL2L2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Antigène BCL-W/BCL2L2
Applications Western Blot
Classification Monoclonal
Clone 90316
Concentration LYOPH
Conjugué Unconjugated
Dilution Western Blot 1 ug/mL
Formule Supplied as a 0.2 μm filtered solution in PBS with Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Numéro d’ordre du gène Q92843
Alias de gène Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2L2, BCL2-like 2, Bclw, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2
Symboles de gène(s) BCL2L2
Espèces hôtes Mouse
Immunogène E. coli-derived recombinant human Bcl-w Met1-Thr172 Accession # Q92843
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 599
Spécificité du test Detects human Bcl-w in Western blots.
Espèces cibles Human
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Type de produit Antibody
Forme Purified
Isotype IgG1
Gamme de produits R&D Systems