Learn More
ACROBiosystems Human beta 2-Microglobulin / B2M Protein, His Tag
Marque: ACROBiosystems B2M-H5225-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
β2 microglobulin is also known as Beta-2-microglobulin (B2M), is a component of MHC class I molecules which belongs to the beta-2-microglobulin family. B2M is present on all nucleated cells (excludes red blood cells). B2M associates not only with the alpha chain of MHC class I molecules, but also with class I-like molecules such as CD1 and Qa. An additional function of B2M is association with the HFE protein, together regulating the expression of hepcidin in the liver which targets the iron transporter ferroportin on the cytoplasmic membrane of enterocytes and macrophages for degradation resulting in decreased iron uptake from food and iron release from recycled red blood cells respectively. Loss of this function causes iron excess and hemochromatosis. Defects in B2M are the cause of hypercatabolic hypoproteinemia (HYCATHYP).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.6 kDa. The protein migrates as 14 kDa under reducing (R) condition (SDS-PAGE).Spécification
AAH32589.1 | |
1 mg | |
Ile 21 - Met 119 | |
B2M | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |
12.6 kDa | |
Human B2M, His Tag (B2M-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (Accession # P61769-1). | |
-20°C | |
beta 2-Microglobulin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |