missing translation for 'onlineSavingsMsg'
                
Learn More
Learn More
Invitrogen™ Human Bisphosphoglycerate mutase (aa 13-112) Control Fragment Recombinant Protein
Code nomenclature Nacres: NA.28
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantité:
                            
                        
                        
                            
                                100 μl
                            
                        
                    
                            
                                Conditionnement: 
                            
                        
                        
                            
                                100µL
                            
                        
                    Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83029 (PA5-83029. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Riboflavin kinase is an essential enzyme that catalyzes the phosphorylation of riboflavin to form flavin mononucleotide, an obligatory step in vitamin B2 utilization and flavin cofactor synthesis.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
        
            
            Spécification
Spécification
| Numéro d’adhésion | P07738 | 
| Concentration | ≥5.0 mg/mL | 
| À utiliser avec (application) | Blocking Assay, Control | 
| Formule | 1 M urea, PBS with no preservative; pH 7.4 | 
| Identification génétique (Entrez) | 669 | 
| Nom | Human Bisphosphoglycerate mutase (aa 13-112) Control Fragment | 
| Quantité | 100 μl | 
| État réglementaire | RUO | 
| Alias de gène | 2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Ab2-098; AI323730; AL022789; Bisphosphoglycerate mutase; BPG-dependent PGAM; Bpgm; C86192; DPGM; erythrocyte 2,3-bisphosphoglycerate mutase; hypothetical protein LOC533785; testis secretory sperm-binding protein Li 202 A; zgc:92230 | 
| Nom usuel | Bisphosphoglycerate mutase | 
| Afficher plus | 
                    Correction du contenu d'un produit
                
                Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
			Invitrogen™ Human Bisphosphoglycerate mutase (aa 13-112) Control Fragment Recombinant Protein > 
            Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu