missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human c-Cbl Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP0423
Code nomenclature Nacres: NA.26
Informations supplémentaires : Poids : 0.01000kg
Description
Cbl Synthetic Peptide, PEP-0423, from Invitrogen.
This peptide corresponds to 14 amino acids near the carboxy terminus of human cbl.PEP-0423 can be used as a blocking peptide with polyclonal antibody PA5-20299.Spécification
867 | |
purified | |
Human c-Cbl | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P22681 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
CBL | |
0.02% sodium azide | |
50 μg | |
PBS |